Skip to Content
Merck
All Photos(1)

Key Documents

409987

Sigma-Aldrich

Di(ethylene glycol) vinyl ether

98%

Synonym(s):

1-Vinyloxy-5-hydroxy-3-oxapentane, 2-[2-(Ethenyloxy)ethoxy]ethanol, 2-[2-(Vinyloxy)ethoxy]ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€83.70
100 ML
€437.00

€83.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€83.70
100 ML
€437.00

About This Item

Linear Formula:
H2C=CH(OCH2CH2)2OH
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€83.70


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.448 (lit.)

bp

196 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

SMILES string

OCCOCCOC=C

InChI

1S/C6H12O3/c1-2-8-5-6-9-4-3-7/h2,7H,1,3-6H2

InChI key

WULAHPYSGCVQHM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service