Skip to Content
Merck
All Photos(1)

Key Documents

409138

Sigma-Aldrich

Poly(dimethylamine-co-epichlorohydrin-co-ethylenediamine) solution

average Mw ~75,000, 50 wt. % in H2O

Synonym(s):

Dimethylamine-ethylenediamine-epichlorohydrin copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mw ~75,000

concentration

50 wt. % in H2O

refractive index

n20/D 1.439

pH

5.0-7.0 (25 °C)

density

1.14 g/mL at 25 °C

SMILES string

CNC.NCCN.ClCC1CO1

InChI

1S/C3H5ClO.C2H8N2.C2H7N/c4-1-3-2-5-3;3-1-2-4;1-3-2/h3H,1-2H2;1-4H2;3H,1-2H3

InChI key

RPKDEWATZPCEIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service