Skip to Content
Merck
All Photos(1)

Key Documents

408468

Sigma-Aldrich

Boc-Asp-OH

99%

Synonym(s):

N-(tert-Butoxycarbonyl)-L-aspartic acid, Boc-L-aspartic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONHCH(COOH)CH2COOH
CAS Number:
Molecular Weight:
233.22
Beilstein:
1913973
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]20/D −6.0°, c = 1 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

116-118 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C9H15NO6/c1-9(2,3)16-8(15)10-5(7(13)14)4-6(11)12/h5H,4H2,1-3H3,(H,10,15)(H,11,12)(H,13,14)/t5-/m0/s1

InChI key

KAJBMCZQVSQJDE-YFKPBYRVSA-N

Gene Information

human ... CA1(759) , CA2(760)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chaitanya R Joshi et al.
Molecular therapy. Nucleic acids, 12, 645-657 (2018-08-07)
Astrocytes, the "star-shaped" glial cells, are appealing gene-delivery targets to treat neurological diseases due to their diverse roles in brain homeostasis and disease. Cationic polymers have successfully delivered genes to mammalian cells and hence present a viable, non-immunogenic alternative to
Mintai P Hwang et al.
Journal of controlled release : official journal of the Controlled Release Society, 318, 270-278 (2019-12-24)
Melanoma is the deadliest type of skin cancer with one of the fastest increasing incidence rates among solid tumors. The use of checkpoint inhibitors (e.g. αPD-1 antibody) has recently emerged as a viable alternative to conventional modes of therapy. However
Yin-Chih Fu et al.
Acta biomaterialia, 10(11), 4583-4596 (2014-07-23)
Nanoparticles (NP) that target bone tissue were developed using PLGA-PEG (poly(lactic-co-glycolic acid)-polyethylene glycol) diblock copolymers and bone-targeting moieties based on aspartic acid, (Asp)(n(1,3)). These NP are expected to enable the transport of hydrophobic drugs. The molecular structures were examined by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service