Skip to Content
Merck
All Photos(2)

Key Documents

406597

Sigma-Aldrich

Poly[(phenyl isocyanate)-co-formaldehyde]

average Mn ~400

Synonym(s):

PMDI, Polymethylene polyphenyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H3(NCO)CH2]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

8.6 (vs air)

form

liquid

specific activity

~3.2 isocyanate groups/molecule

mol wt

average Mn ~400

refractive index

n20/D 1.634

viscosity

1,800 cP(25 °C)(lit.)

bp

392 °C/5 mmHg

density

1.2 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])=O.O=C=Nc1ccccc1

Looking for similar products? Visit Product Comparison Guide

General description

Poly[(phenyl isocyanate)-co-formaldehyde] (P-MDI) is a chain extender with one isocyanate group attached to each of the phenyl groups. It is a liquid polymer that is used to maintain uniformity in the cross-linking reaction between different polymeric units during the curing process. Its crosslinking sites (−NCO) allow it to be utilized in the preparation of polyurethanes.
Poly[(phenyl isocyanate)-co-formaldehyde] (PPIF) is an oligomer that can be used as a chain extender and a cross-linker that forms amide linkages (−NCO). It is highly reactive with hydroxyl groups, which facilitate the formation of a polyurethane network.

Application

P-MDI is an isocyanate that can be used in the preparation of polyurethane resins, films and foams. It can form mechanically robust films which are resistant to solvents and can be used as OTFT based electrodes in biomaterials based applications.
PPIF can be blended with a diol to form a polyurethane mixture with characteristic −NCO peaks. It can also be used as a cross-linker to form high tensile dielectric materials for potential application in organic electronics.

Features and Benefits

High viscosity, high functionality oligomer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Respiratory system, Respiratory system,Eyes,Lungs

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Highly cross-linked polyurethane with enhanced stability of second-order nonlinear optical properties.
Park C, et al.
Macromolecules, 28(10), 3713-3717 (1995)
Polyfunctional active coatings with damage-triggered water-repelling effect.
Latnikova A, et al.
Soft Matter, 7(2), 369-372 (2011)
New thermal study of polymerization and degradation kinetics of methylene diphenyl diisocyanate
da Silva J, et al.
Journal of Thermal Analysis and Calorimetry, 1-8 (2018)
Exploration on high performance polyamide 1010/polyurethane blends filled with functional graphene nanoplatelet: physical properties and technical application
Kausar A
Journal of the Chinese Advanced Materials Society, 5(2), 133-147 (2017)
Second-order nonlinear optical properties and relaxation dynamics of aligned cross-linked polyurethanes with hemicyanine-type chromophores.
Lee K, et al.
Journal of the Optical Society of America B: Optical Physics, 15(1), 393-400 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service