Skip to Content
Merck
All Photos(1)

Documents

397156

Sigma-Aldrich

Pentadecafluorotriethylamine

96%

Synonym(s):

Perfluorotriethylamine, Tris(pentafluoroethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N(CF2CF3)3
CAS Number:
Molecular Weight:
371.05
Beilstein:
1813672
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

2.15 psi ( 20 °C)

Assay

96%

refractive index

n20/D 1.262 (lit.)

bp

68-69 °C/743 mmHg (lit.)

density

1.736 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)N(C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)F

InChI

1S/C6F15N/c7-1(8,9)4(16,17)22(5(18,19)2(10,11)12)6(20,21)3(13,14)15

InChI key

CBEFDCMSEZEGCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentadecafluorotriethylamine (Perfluorotriethylamine, Tris(pentafluoroethyl)amine) is a non-polar organo fluorine compound. Its various physical parameters have been reported. It is an efficient substitute of reaction medium for the Lewis acid mediated reactions. Pyrolysis of perfluorotriethylamine was studied to investigate its thermal behavior in firefighting. Perfluorinated N-ethylpyrrolidine has been identified as the major decomposition product by GC-MS analysis. Production of long-lived microbubbles (50-100μm) in perfluorotriethylamine has been studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition of halon alternatives.
Yamamoto T, et al.
Chemosphere, 35(3), 643-654 (1997)
An efficient high-yield synthesis for perfluorinated tertiary alkyl amines.
Felling KW and Lagow RJ.
Journal of Fluorine Chemistry, 123(2), 233-236 (2003)
Organic reactions without an organic medium. Utilization of perfluorotriethylamine as a reaction medium.
Nakano H and Kitazume T.
Green Chemistry, 1(1), 21-22 (1999)
The behavior of bubbles in perfluorotriethylamine under the effect of strong electric fields.
Korobeinikov SM, et al.
High Temperature, 39(6), 821-825 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service