Skip to Content
Merck
All Photos(2)

Documents

392561

Sigma-Aldrich

Methyl N,N,N′,N′-tetraisopropylphosphorodiamidite

97%

Synonym(s):

Bis(diisopropylamino)methoxyphosphine;

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[(CH3)2CH]2N]2POCH3
CAS Number:
Molecular Weight:
262.37
Beilstein:
4291461
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.461 (lit.)

bp

74-75 °C/0.45 mmHg (lit.)

density

0.915 g/mL at 25 °C (lit.)

SMILES string

COP(N(C(C)C)C(C)C)N(C(C)C)C(C)C

InChI

1S/C13H31N2OP/c1-10(2)14(11(3)4)17(16-9)15(12(5)6)13(7)8/h10-13H,1-9H3

InChI key

YFYBXOIQXOOUCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service