Skip to Content
Merck
All Photos(1)

Key Documents

390712

Sigma-Aldrich

Mer-Trichlorotris(dimethylphenylphosphine)rhenium(III)

97%

Synonym(s):

NSC 168790

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5P(CH3)2]3ReCl3
CAS Number:
Molecular Weight:
707.01
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

core: rhenium
reagent type: catalyst

mp

165 °C (dec.) (lit.)

SMILES string

Cl[Re](Cl)Cl.CP(C)c1ccccc1.CP(C)c2ccccc2.CP(C)c3ccccc3

InChI

1S/3C8H11P.3ClH.Re/c3*1-9(2)8-6-4-3-5-7-8;;;;/h3*3-7H,1-2H3;3*1H;/q;;;;;;+3/p-3

InChI key

OHJDEAZOHPPHHA-UHFFFAOYSA-K

Application

Reactant for synthesis of:
  • Hydridobis(ethylene) and hydrido dinitrogen complexes of rhenium
  • Polyhedral rhenaborane complexes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organometallics, 10, 3105-3105 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service