Skip to Content
Merck
All Photos(1)

Key Documents

385344

Sigma-Aldrich

4-Hydroxy-6-(trifluoromethyl)pyrimidine

99%

Synonym(s):

6-(Trifluoromethyl)-4-pyrimidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3F3N2O
CAS Number:
Molecular Weight:
164.09
Beilstein:
145548
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

169-171 °C (lit.)

functional group

fluoro

SMILES string

Oc1cc(ncn1)C(F)(F)F

InChI

1S/C5H3F3N2O/c6-5(7,8)3-1-4(11)10-2-9-3/h1-2H,(H,9,10,11)

InChI key

LVOYSBZJJWPUBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hydroxy-6-(trifluoromethyl)pyrimidine, a pyrimidine derivative, is a heterocyclic building block.

Application

4-Hydroxy-6-(trifluoromethyl)pyrimidine (6-(Trifluoromethyl)-4-pyrimidinol) may be used in the synthesis of 10-[3-(6-trifluoromethyl-pyrimidin-4-yloxy)-propoxy]-(20S)-camptothecin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingyong Li et al.
Bioorganic & medicinal chemistry, 14(21), 7175-7182 (2006-07-28)
In an attempt to improve the antitumor activity and decrease the cytotoxicity of camptothecin, 18 new 10-substituted camptothecin derivatives were prepared. The cytotoxicity in vitro on cancer cell lines and antitumor activity in vivo, and inhibitory properties of topoisomerase I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service