Skip to Content
Merck
All Photos(1)

Key Documents

374903

Sigma-Aldrich

2,5-Difluorophenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NCO
CAS Number:
Molecular Weight:
155.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.494 (lit.)

density

1.323 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1ccc(F)c(c1)N=C=O

InChI

1S/C7H3F2NO/c8-5-1-2-6(9)7(3-5)10-4-11/h1-3H

InChI key

SNHIIFOXCRYGGY-UHFFFAOYSA-N

General description

2,5-Difluorophenyl isocyanate is an aryl fluorinated building block. It is also referred as 2,5-difluorophenyl ester of isocyanic acid.

Application

2,5-Difluorophenyl isocyanate may be used in the preparation of a potential antiatherosclerotic agent.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V G DeVries et al.
Journal of medicinal chemistry, 32(10), 2318-2325 (1989-10-01)
The discovery that a series of N,N-dialkyl-N'-arylureas were inhibitors of the ACAT enzyme has led to a structure-activity study involving the systematic modification of three sites of the urea backbone. This study culminated in the selection of N'-(2,4-dimethylphenyl)-N-benzyl-N-n-butylurea (115) for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service