Skip to Content
Merck
All Photos(2)

Key Documents

374113

Sigma-Aldrich

(S)-(+)-5-(Trityloxymethyl)-2-pyrrolidinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23NO2
CAS Number:
Molecular Weight:
357.44
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +15°, c = 1 in chloroform

mp

164-166 °C (lit.)

functional group

ether
phenyl

SMILES string

O=C1CC[C@@H](COC(c2ccccc2)(c3ccccc3)c4ccccc4)N1

InChI

1S/C24H23NO2/c26-23-17-16-22(25-23)18-27-24(19-10-4-1-5-11-19,20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-15,22H,16-18H2,(H,25,26)/t22-/m0/s1

InChI key

FQSSRLVFPQIRJK-QFIPXVFZSA-N

Application

Used in asymmetric Diels-Alder reactions and asymmetric conjugate additions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 426-426 (1990)
Journal of the Chemical Society. Chemical Communications, 1177-1177 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service