Skip to Content
Merck
All Photos(1)

Key Documents

364045

Sigma-Aldrich

Ethyl-1-propenyl ether, mixture of cis and trans

98%

Synonym(s):

1-Ethoxypropene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH=CHCH3
CAS Number:
Molecular Weight:
86.13
Beilstein:
1735974
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

refractive index

n20/D 1.398 (lit.)

bp

67-76 °C (lit.)

density

0.778 g/mL at 25 °C (lit.)

SMILES string

CCO\C=C\C

InChI

1S/C5H10O/c1-3-5-6-4-2/h3,5H,4H2,1-2H3/b5-3+

InChI key

XDHOEHJVXXTEDV-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service