Skip to Content
Merck
All Photos(1)

Documents

360333

Sigma-Aldrich

Sodium phosphonoformate tribasic hexahydrate

98%

Synonym(s):

Foscarnet, Phosphonoformic acid trisodium salt hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O3PCO2Na · 6H2O
CAS Number:
Molecular Weight:
300.04
MDL number:
PubChem Substance ID:

Assay

98%

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O

InChI

1S/CH3O5P.3Na.6H2O/c2-1(3)7(4,5)6;;;;;;;;;/h(H,2,3)(H2,4,5,6);;;;6*1H2/q;3*+1;;;;;;/p-3

InChI key

ILRVASBWNRYBFD-UHFFFAOYSA-K


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jocelyne Piret et al.
Journal of virology, 89(8), 4636-4644 (2015-02-13)
DNA polymerases of the Herpesviridae and bacteriophage RB69 belong to the α-like DNA polymerase family. In spite of similarities in structure and function, the RB69 enzyme is relatively resistant to foscarnet, requiring the mutation V478W in helix N to promote

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service