Skip to Content
Merck
All Photos(1)

Documents

340308

Sigma-Aldrich

(R)-(−)-2-Hexanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718998
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

99%

optical activity

[α]24/D −11°, neat

refractive index

n20/D 1.414 (lit.)

bp

137-138 °C (lit.)

density

0.814 g/mL at 20 °C (lit.)

SMILES string

CCCC[C@@H](C)O

InChI

1S/C6H14O/c1-3-4-5-6(2)7/h6-7H,3-5H2,1-2H3/t6-/m1/s1

InChI key

QNVRIHYSUZMSGM-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of key intermediates for model studies in the total synthesis of antivirally active glycolipid cycloviracin B1.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fangyuan Zhou et al.
The ISME journal, 11(12), 2809-2820 (2017-08-12)
Interactions among microbial symbionts have multiple roles in the maintenance of insect-microbe symbiosis. However, signals mediating microbial interactions have been scarcely studied. In the classical model system of bark beetles and fungal associates, fungi increase the fitness of insects. However
Kazumi Kondo et al.
Journal of medicinal chemistry, 45(17), 3805-3808 (2002-08-09)
The synthesis and evaluation of both the (R)- and (S)-enantioisomers about the 5-position on a tetrahydro-1H-1-benzazepine derivative were described. The absolute configuration of the (R,R)-isomer (10) was determined by X-ray crystallographic analysis. After evaluation of both enantiomers (compounds R-2, S-2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service