Skip to Content
Merck
All Photos(1)

Key Documents

339008

Sigma-Aldrich

Ammonium-d4 dideuteriumphosphate

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ND4)D2PO4
CAS Number:
Molecular Weight:
121.06
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

99 atom % D

Quality Level

mp

195-197 °C (lit.)

mass shift

M+6

SMILES string

[2H]N([2H])[2H].[2H]OP(=O)(O[2H])O[2H]


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service