Skip to Content
Merck
All Photos(2)

Key Documents

336688

Sigma-Aldrich

Potassium hexafluorozirconate

Synonym(s):

Dipotassium hexafluorozirconate, Dipotassium zirconium hexafluoride, Potassium fluorozirconate, Potassium fluorozirconate (K2 ZrF6 ), Potassium hexafluorozirconate (K2 ZrF6 ), Potassium hexafluorozirconate(IV), Potassium zirconium fluoride (K2 ZrF6 ), Potassium zirconium hexafluoride, Zirconium potassium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2ZrF6
CAS Number:
Molecular Weight:
283.41
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

SMILES string

[K+].[K+].F[Zr--](F)(F)(F)(F)F

InChI

1S/6FH.2K.Zr/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

BJZIJOLEWHWTJO-UHFFFAOYSA-H

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service