Skip to Content
Merck
All Photos(1)

Documents

336645

Sigma-Aldrich

Potassium heptafluoroniobate(V)

98%

Synonym(s):

Dipotassium heptafluoroniobate, Niobium potassium fluoride (NbK2 F7 ), Potassium niobium fluoride(K2NbF7)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2NbF7
CAS Number:
Molecular Weight:
304.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

98%

form

powder

SMILES string

[K+].[K+].F[Nb--](F)(F)(F)(F)(F)F

InChI

1S/7FH.2K.Nb/h7*1H;;;/q;;;;;;;2*+1;+5/p-7

InChI key

FTPUNAWAGWERLA-UHFFFAOYSA-G

Looking for similar products? Visit Product Comparison Guide

Application

Used in the electrochemical synthesis of NbB2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chem. Abstr., 122, 18820q-18820q (1995)
Deviatkin, S.V. et al.
Proc. Electrochem. Soc., 93-9, 584-584 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service