Skip to Content
Merck
All Photos(1)

Key Documents

307688

Sigma-Aldrich

Cerium(III) sulfate

97%

Synonym(s):

Cerium sesquisulfate, Cerous sulfate, Dicerium trisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce2(SO4)3
CAS Number:
Molecular Weight:
568.42
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

reagent type: catalyst
core: cerium

SMILES string

[Ce+3].[Ce+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Ce.3H2O4S/c;;3*1-5(2,3)4/h;;3*(H2,1,2,3,4)/q2*+3;;;/p-6

InChI key

OZECDDHOAMNMQI-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammar Bader et al.
Phytochemistry, 116, 262-268 (2015-04-22)
Two flavonoids, jaceosidin 7-O-β-d-glucopyranosyl-(1→2)-β-d-glucopyranoside (1) and hispidulin 7-O-β-d-glucopyranosyl-(1→2)-β-d-glucopyranoside (2), and one phenylpropanoid, 3,3'-dimethyl-lunariifolioside (3), along with 11 known compounds (4-14), were isolated from the aerial parts of Phlomis kurdica growing in Jordan. Structures of 1-3 were elucidated on the basis
Xiaoming Chen et al.
Nature communications, 6, 7204-7204 (2015-05-23)
The structurally complex alkaloid gelsemine was previously thought to have no significant biological activities, but a recent study has shown that it has potent and specific antinociception in chronic pain. While this molecule has attracted significant interests from the synthetic
Nilasha Banerjee et al.
Nuclear medicine and biology, 42(3), 274-282 (2014-12-30)
Organic Anion Transporting Polypeptides (OATP) are a family of membrane associated transporters that facilitate estrone-3-sulphate (E3S) uptake by hormone dependent, post-menopausal breast cancers. We have established E3S as a potential ligand for targeting hormone dependent breast cancer cells, and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service