Skip to Content
Merck
All Photos(1)

Key Documents

29177

Sigma-Aldrich

(S)-4-Chloro-1,2-epoxybutane

≥98.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7ClO
CAS Number:
Molecular Weight:
106.55
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98.0% (sum of enantiomers, GC)

optical purity

enantiomeric ratio: ≥98.5:1.5 (GC)

SMILES string

ClCC[C@H]1CO1

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service