All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5F6O3
CAS Number:
Molecular Weight:
222.04
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
97%
refractive index
n20/D 1.324 (lit.)
bp
72 °C (lit.)
density
1.654 g/mL at 25 °C (lit.)
SMILES string
FC1(F)C(=O)OC(=O)C(F)(F)C1(F)F
InChI
1S/C5F6O3/c6-3(7)1(12)14-2(13)4(8,9)5(3,10)11
InChI key
IHYAGCYJVNHXCT-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service