Skip to Content
Merck
All Photos(1)

Key Documents

277886

Sigma-Aldrich

6-Chloro-3,5-diamino-2-pyrazinecarboxamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6ClN5O
CAS Number:
Molecular Weight:
187.59
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

98%

form

solid

mp

219-221 °C (lit.)

SMILES string

NC(=O)c1nc(Cl)c(N)nc1N

InChI

1S/C5H6ClN5O/c6-2-4(8)11-3(7)1(10-2)5(9)12/h(H2,9,12)(H4,7,8,11)

InChI key

UYTQEWLAYXCJEC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Alvarez de la Rosa et al.
The Journal of general physiology, 124(4), 395-407 (2004-09-29)
The serum and glucocorticoid induced kinase 1 (SGK1) participates in the regulation of sodium reabsorption in the distal segment of the renal tubule, where it may modify the function of the epithelial sodium channel (ENaC). The molecular mechanism underlying SGK1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service