Skip to Content
Merck
All Photos(1)

Documents

275387

Sigma-Aldrich

3-Fluorothiophenol

95%

Synonym(s):

3-Fluorobenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SH
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

bp

170 °C (lit.)

density

1.517 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(S)c1

InChI

1S/C6H5FS/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

ZDEUGINAVLMAET-UHFFFAOYSA-N

Application

3-Fluorothiophenol was used in the synthesis of dibenzo[bc,fg]dithiapentalene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient protocol for synthesizing dibenzodithiapentalenes.
Tetrahedron Letters, 52(31), 4045-4047 (2011)
Javier Campanini-Salinas et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-22)
A rapid emergence of resistant bacteria is occurring worldwide, endangering the efficacy of antibiotics and reducing the therapeutic arsenal available for treatment of infectious diseases. In the present study, we developed a new class of compounds with antibacterial activity obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service