Skip to Content
Merck
All Photos(2)

Key Documents

269425

Sigma-Aldrich

4-Nitrophenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NCO
CAS Number:
Molecular Weight:
164.12
Beilstein:
389516
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

137-138 °C/11 mmHg (lit.)

mp

56-59 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(cc1)N=C=O

InChI

1S/C7H4N2O3/c10-5-8-6-1-3-7(4-2-6)9(11)12/h1-4H

InChI key

GFNKTLQTQSALEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl isocyanate can be used as a reactant to synthesize:
  • 4-aminophenyl carbamic acid methyl ester by Raney nickel catalyzed hydrogenation in methanol-DCM solvent system.[1]
  • Urea-functionalized derivatives with p-nitrobenzene units by reacting with 1,2-bis(2′-aminophenoxy) benzene.[2]
  • 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide by treating with 5-methyl-3,4-diphenyl-1H-pyrrole-2-carbohydrazide.[3]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Crystal Structure and Anion Recognition Property of a Flexible Urea-functionalized Receptor
Chao H, et al.
Chinese Journal of Structural Chemistry / Jie Gou Hua Xue, (05), 737-744 (2019)
Pyrrolylamidourea based anion receptors
Evans LS, et al.
New. J. Chem., 30(7), 1019-1025 (2006)
Pyrrolylamidourea based anion receptors.
Evans LS, et al.
New. J. Chem., 30(7), 1019-1025 (2006)
Novel and Efficient One-Pot Synthesis of (aminophenyl) Carbamic Acid Esters
Garofalo A, et al.
Synthetic Communications, 41(13), 2007-2016 (2011)
Junta Sano et al.
Polymers, 12(5) (2020-05-24)
Novel interpenetrating polymer networks (IPNs) were synthesized from N-isopropylacrylamide (NIPAM) and polysiloxanes containing a urea or thiourea side group, in addition to the silanol residue, through two reactions, such as the radical gelation of NIPAM and the condensation of silanols

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service