Skip to Content
Merck
All Photos(3)

Key Documents

262552

Sigma-Aldrich

Potassium fluoride on Celite®

extent of labeling: 50 wt. % loading, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF-Celite
Molecular Weight:
58.10
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
technical grade
form:
powder and chunks

grade

technical grade

form

powder and chunks

extent of labeling

50 wt. % loading

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel L Camp et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(28), 8690-8695 (2015-06-24)
Most aquatic vertebrates use suction to capture food, relying on rapid expansion of the mouth cavity to accelerate water and food into the mouth. In ray-finned fishes, mouth expansion is both fast and forceful, and therefore requires considerable power. However
Henry C Astley et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(19), 6200-6205 (2015-04-02)
Many organisms move using traveling waves of body undulation, and most work has focused on single-plane undulations in fluids. Less attention has been paid to multiplane undulations, which are particularly important in terrestrial environments where vertical undulations can regulate substrate
Marianna Mamusa et al.
Faraday discussions, 181, 193-209 (2015-05-12)
Maghemite (γ-Fe2O3) nanoparticles (NPs) can be successfully dispersed in a protic ionic liquid, ethylammonium nitrate (EAN), by transfer from aqueous dispersions into EAN. As the aqueous systems are well controlled, several parameters can be tuned. Their crucial role towards the
Pierre Czernic et al.
Plant physiology, 169(2), 1254-1265 (2015-08-20)
Nutritional symbiotic interactions require the housing of large numbers of microbial symbionts, which produce essential compounds for the growth of the host. In the legume-rhizobium nitrogen-fixing symbiosis, thousands of rhizobium microsymbionts, called bacteroids, are confined intracellularly within highly specialized symbiotic
W De Jaeghere et al.
International journal of pharmaceutics, 492(1-2), 1-9 (2015-07-15)
The primary purpose of this study was to process partially hydrolyzed PVOH grades (degree of hydroxylation (DH): 33-88%) via HME and to evaluate them as carrier for oral immediate release dosage forms in order to improve the release rate of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service