250104
1-Bromo-2,3,5,6-tetramethylbenzene
98%
Synonym(s):
3-Bromodurene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HC6(CH3)4Br
CAS Number:
Molecular Weight:
213.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
98%
bp
111-113 °C/6 mmHg (lit.)
mp
60-62 °C (lit.)
SMILES string
Cc1cc(C)c(C)c(Br)c1C
InChI
1S/C10H13Br/c1-6-5-7(2)9(4)10(11)8(6)3/h5H,1-4H3
InChI key
WJKBPTLQJXKEHC-UHFFFAOYSA-N
Application
1-Bromo-2,3,5,6-tetramethylbenzene has been used in the preparation of:
- 1-(3′,5′-bis(trifluoromethyl)phenyl)pyromellitic dianhydride, mono-substituted dianhydride monomer
- 2-(N,N-dimethylamino)phenylbis(2,3,5,6-tetramethylphenyl)borane
- 2-(N,N-diethylamino)phenyl-2,3,5,6-tetramethylphenylborinic acid
- 3-(2,3,5,6-tetramethylphenyl)acrylamide.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and properties of fluorescent organoboranes: triarylmethane-type dyes.
Albrecht K, et al.
J. Chem. Soc. Perkin Trans. II, 2153-2157 (2000)
Synthesis and characterization of polyimides from novel 1-(3', 5'-bis (trifluoromethyl) benzene) pyromelliticdianhydride (6FPPMDA).
Myung BY, et al.
Polymer, 45(10), 3185-3193 (2004)
Synthesis of 3-arylpropylamine derivatives from aryl halides using Heck reaction.
Baek GH, et al.
Bull. Korean Chem. Soc., 20(2), 232-236 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service