Skip to Content
Merck
All Photos(1)

Documents

239453

Sigma-Aldrich

Methyl chloroacetate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COOCH3
CAS Number:
Molecular Weight:
108.52
Beilstein:
506255
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

3.8 (vs air)

vapor pressure

10 mmHg ( 25 °C)
5.25 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

869 °F

expl. lim.

18.5 %

refractive index

n20/D 1.422 (lit.)

bp

130 °C/740 mmHg (lit.)

mp

−33 °C (lit.)

density

1.238 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCl

InChI

1S/C3H5ClO2/c1-6-3(5)2-4/h2H2,1H3

InChI key

QABLOFMHHSOFRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service