Skip to Content
Merck
All Photos(1)

Key Documents

232947

Sigma-Aldrich

2-(Trifluoromethyl)benzonitrile

98%

Synonym(s):

α,α,α-Trifluoro-o-tolunitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CN
CAS Number:
Molecular Weight:
171.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.4632 (lit.)

mp

7.5 °C (lit.)

density

1.294 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccccc1C#N

InChI

1S/C8H4F3N/c9-8(10,11)7-4-2-1-3-6(7)5-12/h1-4H

InChI key

SOZGHDCEWOLLHV-UHFFFAOYSA-N

General description

2-(Trifluoromethyl)benzonitrile reacts with tert-butyl acetate in the presence of sulfuric acid to give the corresponding N-tert-butyl amides[1].

Application

2-(Trifluoromethyl)benzonitrile was used in the synthesis of symmetrical N,N′-alkylidine bisamides[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient and efficient protocol for the synthesis of symmetrical N,N'-alkylidine bisamides by sulfamic acid under solvent-free conditions.
Selvam NP, et al.
Canadian Journal of Chemistry, 86(1), 32-38 (2008)
An efficient method for the conversion of aromatic and aliphatic nitriles to the corresponding N-tert-butyl amides: a modified Ritter reaction.
Reddy KL.
Tetrahedron Letters, 44(7), 1453-1455 (2003)
Megumi Morimoto et al.
PloS one, 12(12), e0189480-e0189480 (2017-12-08)
Sarcopenia and cachexia present characteristic features of a decrease in skeletal muscle mass and strength, anorexia, and lack of motivation. Treatments for these diseases have not yet been established, although selective androgen receptor modulators (SARMs) are considered as therapeutic targets.
Nobuyuki Ishikura et al.
International journal of oncology, 46(4), 1560-1572 (2015-01-31)
Resistance of prostate cancer to castration is currently an unavoidable problem. The major mechanisms underlying such resistance are androgen receptor (AR) overexpression, androgen-independent activation of AR, and AR mutation. To address this problem, we developed an AR pure antagonist, CH5137291

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service