Skip to Content
Merck
All Photos(1)

Key Documents

230960

Sigma-Aldrich

Stains-All

Dye content 95 %

Synonym(s):

1-Ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide, 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H27BrN2S2
CAS Number:
Molecular Weight:
559.58
Beilstein:
3865213
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 95%

λmax

597 nm

SMILES string

[Br-].CCN1C(/Sc2ccc3ccccc3c12)=C\C(C)=C\c4sc5ccc6ccccc6c5[n+]4CC

InChI

1S/C30H27N2S2.BrH/c1-4-31-27(33-25-16-14-21-10-6-8-12-23(21)29(25)31)18-20(3)19-28-32(5-2)30-24-13-9-7-11-22(24)15-17-26(30)34-28;/h6-19H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

MPBRYMWMMKKRGC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Krystyna Kosinska et al.
PloS one, 10(5), e0125192-e0125192 (2015-05-02)
Hyaluronic acid (HA), lubricin, and phospholipid species (PLs) contribute independently or together to the boundary lubrication of articular joints that is provided by synovial fluid (SF). Our study is the first reporting quantitative data about the molecular weight (MW) forms
M Vilanova et al.
The Biochemical journal, 251(3), 901-905 (1988-05-01)
The isolated activation segment of pig procarboxypeptidase A binds two Tb3+ ions in a strong and specific way. In contrast, the binding of Ca2+, Cd2+ and Mg2+ is weak. The binding of Tb3+ increases the resistance of the isolated activation
M A Hernández et al.
Biochimica et biophysica acta, 965(2-3), 195-201 (1988-05-12)
Calcium has been suggested to be an important element in the regulation of microtubule dynamics 'in vivo'. In this report we have analyzed the possibility that microtubule-associated protein 2 (MAP2) binds calcium. MAP2 was blue-stained with the cationic carbocyanine dye
Interaction of drugs with branched DNA structures.
M Lu et al.
Critical reviews in biochemistry and molecular biology, 27(3), 157-190 (1992-01-01)
P C Denny et al.
Journal of dental research, 70(10), 1320-1327 (1991-10-01)
The predominant mucins in human whole saliva, MG1 and MG2, serve to protect and to lubricate the oral cavity. In this study, both unstimulated and stimulated whole salivas were collected from two groups of subjects: young (18-35 years of age)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service