Skip to Content
Merck
All Photos(1)

Documents

226335

Sigma-Aldrich

Bis(methylthio)methane

99%

Synonym(s):

Dimethylthiomethane, Formaldehyde dimethyl mercaptal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2SCH3
CAS Number:
Molecular Weight:
108.23
Beilstein:
1731143
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.53 (lit.)

bp

147 °C (lit.)

solubility

aqueous base: insoluble(lit.)

density

1.059 g/mL at 25 °C (lit.)

SMILES string

CSCSC

InChI

1S/C3H8S2/c1-4-3-5-2/h3H2,1-2H3

InChI key

LOCDPORVFVOGCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The liquid structure of bis(methylthio)methane at room temperature was studied by the bonding and non-bonding interatomic potential functions (FMP-RMC) simulation.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orsolya Gereben et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(45), 454201-454201 (2013-10-22)
Series of flexible molecule reverse Monte Carlo calculations, using bonding and non-bonding interatomic potential functions (FMP-RMC), were performed starting from previous molecular dynamics results that had applied the OPLS-AA and EncadS force fields. During RMC modeling, the experimental x-ray total
Osvaldo Zarivi et al.
FEMS microbiology letters, 220(1), 81-88 (2003-03-20)
Tyrosinase is an enzyme having two copper atoms at the reactive site occurring in prokaryotic and eukaryotic organisms. In animals tyrosinase is responsible for pigmentation, in plants for protection of injured tissues or, as in fungi, to harden cell walls.
Mohamed Osman Awaleh et al.
Inorganic chemistry, 46(8), 3185-3191 (2007-03-17)
Two topologically comparable complexes, [Ag6(CF3CO2)3(L1-Me)3(SCH3)3]infinity (1) and [Ag6(CF3CF2CO2)3(L1-Me)2(SCH3)3(H2O)]infinity (2), were prepared and characterized by single-crystal diffractometry. The structures consist of Ag12S6 clusters linked by bis(methylthio)methane ligands, L1-Me, thus forming 1D coordination polymers. The 12 Ag atoms of the cluster are
Pascal Fuchsmann et al.
Journal of agricultural and food chemistry, 63(34), 7511-7521 (2015-08-01)
To establish the odor profiles of three differently fabricated commercial Swiss Tilsit cheeses, analyses were conducted using headspace solid-phase microextraction gas chromatography-mass spectrometry/pulsed flame photometric detection and gas chromatography-olfactometry to identify and quantitate volatile compounds. In addition, odor quality and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service