Skip to Content
Merck
All Photos(1)

Key Documents

225703

Sigma-Aldrich

3-Chloro-2,2-dimethylpropionyl chloride

98%

Synonym(s):

β-Chloropivaloyl chloride, 3-Chloropivaloyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2C(CH3)2COCl
CAS Number:
Molecular Weight:
154.01
Beilstein:
1746729
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
liquid
Assay:
98%
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

85-86 °C/60 mmHg (lit.)

density

1.199 g/mL at 25 °C (lit.)

SMILES string

CC(C)(CCl)C(Cl)=O

InChI

1S/C5H8Cl2O/c1-5(2,3-6)4(7)8/h3H2,1-2H3

InChI key

MQZNDDUMJVSIMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chloro-2,2-dimethylpropionyl chloride may be used in chemical synthesis studies.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service