Skip to Content
Merck
All Photos(2)

Key Documents

221597

Sigma-Aldrich

3,4-Diethoxybenzoic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2C6H3CO2H
CAS Number:
Molecular Weight:
210.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

167-169 °C (lit.)

functional group

carboxylic acid

SMILES string

CCOc1ccc(cc1OCC)C(O)=O

InChI

1S/C11H14O4/c1-3-14-9-6-5-8(11(12)13)7-10(9)15-4-2/h5-7H,3-4H2,1-2H3,(H,12,13)

InChI key

VVKCVAPLTRZJHH-UHFFFAOYSA-N

Application

3,4-Diethoxybenzoic acid was used in the synthesis of dimethyl 2-(3,4-diethoxyphenyI)-2-methylmalonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymes in organic synthesis 51. Probing the dimensions of the large hydrophobic pocket of the active site of pig liver esterase.
Tetrahedron Asymmetry, 4(9), 2025-2040 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service