Skip to Content
Merck
All Photos(2)

Key Documents

220817

Sigma-Aldrich

2-(Trimethylsilyl)-1,3-dithiane

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16S2Si
CAS Number:
Molecular Weight:
192.42
Beilstein:
1616463
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

refractive index

n20/D 1.533 (lit.)

bp

54-55 °C/0.17 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

C[Si](C)(C)C1SCCCS1

InChI

1S/C7H16S2Si/c1-10(2,3)7-8-5-4-6-9-7/h7H,4-6H2,1-3H3

InChI key

BTTUMVHWIAXYPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Trimethylsilyl)-1,3-dithiane participates in Lewis base-catalyzed 1,3-dithiane addition to electrophiles such as carbonyl compounds and N-substituted aldimines[1]. It undergoes novel diazo transfer reaction with tosyl azide in hexamethylphosphoramide-THF to yield 2-diazo-1,3-dithiane, which on decomposition yields formal carbene adducts[2]. It is a versatile acyl anion equivalent[3][4].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diazo transfer reaction of 2-(trimethylsilyl)-1, 3-dithiane with tosyl azide. Carbenic reactivity of transient 2-diazo-1, 3-dithiane.
Benati L, et al.
Tetrahedron, 53(27), 9269-9278 (1997)
Smith, A. B., III; Boldi, A. M.
Journal of the American Chemical Society, 119, 6925-6925 (1997)
Makoto Michida et al.
Chemistry, an Asian journal, 3(8-9), 1592-1600 (2008-06-21)
Lewis base-catalyzed 1,3-dithiane addition to electrophiles such as carbonyl compounds and N-substituted aldimines with 2-trimethylsilyl-1,3-dithiane (TMS-dithiane) is described. By the activation of the carbon-silicon bond in the presence of a Lewis base catalyst such as tetrabutylammonium phenoxide (PhONnBu(4)), a 1,3-dithiane
Amos B Smith et al.
Journal of the American Chemical Society, 125(47), 14435-14445 (2003-11-20)
The development, application, and advantages of a one-flask multicomponent dithiane linchpin coupling protocol, over the more conventional stepwise addition of dithiane anions to electrophiles leading to the rapid, efficient, and stereocontrolled assembly of highly functionalized intermediates for complex molecule synthesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service