Skip to Content
Merck
All Photos(2)

Key Documents

218529

Sigma-Aldrich

4-Morpholinecarbonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N2O
CAS Number:
Molecular Weight:
112.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.473 (lit.)

bp

71-73 °C/0.6 mmHg (lit.)

density

1.109 g/mL at 25 °C (lit.)

SMILES string

N#CN1CCOCC1

InChI

1S/C5H8N2O/c6-5-7-1-3-8-4-2-7/h1-4H2

InChI key

BOQOXLAQTDFJKU-UHFFFAOYSA-N

General description

He I and He II photoelectron spectra of 4-morpholinecarbonitrile has been investigated[1].

Application

4-Morpholinecarbonitrile was used in the synthesis of substituted heterocyclic compounds[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tautomerie von Heterocyclen, VII. Synthese und Reaktionen N-vinylsubstituierter Chlorformamidine.
Ried W and Erle H-E.
Chemische Berichte, 112(2), 640-647 (1979)
Trans-annular interactions in morpholines: A photoelectron spectroscopic study.
Novak I and Kovac B.
Heteroatom Chem., 6(1), 29-31 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service