Skip to Content
Merck
All Photos(1)

Key Documents

203785

Sigma-Aldrich

Mercury(II) iodide

99.99% trace metals basis

Synonym(s):

Mercuric iodide red

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgI2
CAS Number:
Molecular Weight:
454.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: mercury

impurities

≤150.0 ppm Trace Metal Analysis

bp

354 °C (lit.)

mp

259 °C (lit.)

solubility

H2O: soluble

SMILES string

I[Hg]I

InChI

1S/Hg.2HI/h;2*1H/q+2;;/p-2

InChI key

YFDLHELOZYVNJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Health hazard from mercury soap.
E S Mucklow
Lancet (London, England), 1(8635), 448-448 (1989-02-25)
Zhong Su et al.
Physics in medicine and biology, 50(12), 2907-2928 (2005-06-03)
The signal properties of polycrystalline mercuric iodide (HgI2) film detectors, under irradiation conditions relevant to mammographic, radiographic, fluoroscopic and radiotherapy x-ray imaging, are reported. Each film detector consists of an approximately 230 to approximately 460 microm thick layer of HgI2
N C Sukul et al.
Homeopathy : the journal of the Faculty of Homeopathy, 91(4), 217-220 (2002-11-09)
Mercuric chloride 30c and Mercuric iodide 30c were prepared by successive dilution in 30 steps of 1:100 followed by sonication at 20KHz for 30s at each step. Both were prepared in two media: 90% ethanol and distilled water. Three preparations
Acute renal failure associated with application of a mercuric blister in a horse.
M D Markel et al.
Journal of the American Veterinary Medical Association, 185(1), 92-94 (1984-07-01)
H B Barber et al.
Medical physics, 18(3), 373-381 (1991-05-01)
The physical properties of three surgical probes containing different radiation detectors are compared: a NaI(Tl) scintillator with a flexible, fiber-optic light guide, and two semiconductor detectors that operate at room temperature, CdTe and HgI2. Also compared are spectra, energy resolutions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service