Skip to Content
Merck
All Photos(1)

Documents

189391

Sigma-Aldrich

Poly(1-butene), isotactic

average Mw ~570,000 by GPC, pellets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C2H5)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

pellets

melt index

0.4

mol wt

average Mw ~570,000 by GPC

transition temp

Tg −24 °C
Tm 125 °C

density

0.91 g/mL at 25 °C (lit.)

SMILES string

CCC=C

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service