Skip to Content
Merck
All Photos(1)

Key Documents

18886

Sigma-Aldrich

2,4-Pentadienoic acid

≥97.0% (T)

Synonym(s):

1,3-Butadiene-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH=CHCOOH
CAS Number:
Molecular Weight:
98.10
Beilstein:
1739248
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (T)

form

solid

contains

hydroquinone as stabilizer

mp

69-72 °C

solubility

1 M NaOH: soluble 0.5 g/10 mL, clear, brown

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

OC(=O)\C=C\C=C

InChI

1S/C5H6O2/c1-2-3-4-5(6)7/h2-4H,1H2,(H,6,7)/b4-3+

InChI key

SDVVLIIVFBKBMG-ONEGZZNKSA-N

Application

2,4-Pentadienoic acid was used in preparation of chiral propargylester[1]. It was also used in the preparation of trans 1-N-acylamino-1,3-dienes via modified Curtius procedure[2].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

trans-1-N-Acylamino-1, 3-dienes: preparation from dienoic acids.
Overman L, et al.
The Journal of Organic Chemistry, 43(11), 2164-2167 (1978)
Xiaohu Yi et al.
ChemSusChem, 10(7), 1494-1500 (2017-01-18)
A series of choline (Ch)-exchanged heteropoly acids (HOCH
Daichi Oguro et al.
Bioscience, biotechnology, and biochemistry, 75(8), 1502-1505 (2011-08-09)
The synthesis and sensory evaluation of enantiomeric sets of sedanenolide (1) and 3-butylphthalide (3) are described. The asymmetric synthesis was achieved via the intramolecular Diels-Alder reaction of chiral propargylester (5) which was prepared from optically active propargyl alcohol (4) and
Brian D Hudson et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(12), 4951-4965 (2012-08-25)
When it is difficult to develop selective ligands within a family of related G-protein-coupled receptors (GPCRs), chemically engineered receptors activated solely by synthetic ligands (RASSLs) are useful alternatives for probing receptor function. In the present work, we explored whether a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service