Skip to Content
Merck
All Photos(1)

Key Documents

17752

Sigma-Aldrich

N1-Boc-2,2′-iminodiethylamine

≥97.0%

Synonym(s):

N1-Boc-diethylenetriamine, tert-Butyl [2-(2-aminoethylamino)ethyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21N3O2
CAS Number:
Molecular Weight:
203.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0%

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.474

density

1.020 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)OC(=O)NCCNCCN

InChI

1S/C9H21N3O2/c1-9(2,3)14-8(13)12-7-6-11-5-4-10/h11H,4-7,10H2,1-3H3,(H,12,13)

InChI key

KQXGYFNJIMVFDT-UHFFFAOYSA-N

Application

Protected building block for synthesizing scaffolds for combinatorial chemistry; building block for polyamines, chelators etc [1][2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Adamczyk et al.
Organic preparations and procedures international, 30, 339-339 (1998)
A.P. Krapcho et al.
Synthetic Communications, 20, 2559-2559 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service