Skip to Content
Merck
All Photos(1)

Documents

172049

Sigma-Aldrich

2-Hydroxyethyl salicylate

97%

Synonym(s):

Ethylene glycol monosalicylate, Ethylene glycol salicylate, Salicylic acid 2-hydroxyethyl ester, Salicylic acid ethylene glycol ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COOCH2CH2OH
CAS Number:
Molecular Weight:
182.17
Beilstein:
2255685
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.548 (lit.)

bp

166 °C/13 mmHg (lit.)

density

1.244 g/mL at 25 °C (lit.)

SMILES string

OCCOC(=O)c1ccccc1O

InChI

1S/C9H10O4/c10-5-6-13-9(12)7-3-1-2-4-8(7)11/h1-4,10-11H,5-6H2

InChI key

LVYLCBNXHHHPSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service