Skip to Content
Merck
All Photos(1)

Key Documents

151041

Sigma-Aldrich

2,4-Dimethylbenzaldehyde

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CHO
CAS Number:
Molecular Weight:
134.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.549 (lit.)

bp

102-103 °C/14 mmHg (lit.)

mp

−9 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

Cc1ccc(C=O)c(C)c1

InChI

1S/C9H10O/c1-7-3-4-9(6-10)8(2)5-7/h3-6H,1-2H3

InChI key

GISVICWQYMUPJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The D values of 2,4-Dimethylbenzaldehyde were studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determinations of large ZFS D-parameters in 2,4-, 2,5- and 3,4-dimethybenzaldehydes dispersed in durene single crystals.
Despres A and Migirdicyan E.
Chemical Physics, 22(1), 121-130 (1977)
Matthias Onghena et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 31(12), 2090-2102 (2014-11-20)
In 2011, the European Union prohibited the production of polycarbonate (PC) baby bottles due to the toxic effects of the PC monomer bisphenol-A. Therefore, baby bottles made of alternative materials, e.g. polypropylene (PP) or polyethersulphone (PES), are currently marketed. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service