Skip to Content
Merck
All Photos(1)

Documents

150576

Sigma-Aldrich

5-Norbornene-2-carbonitrile, mixture of isomers

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N
CAS Number:
Molecular Weight:
119.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

82-86 °C/10 mmHg (lit.)

mp

12-14 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

N#CC1CC2CC1C=C2

InChI

1S/C8H9N/c9-5-8-4-6-1-2-7(8)3-6/h1-2,6-8H,3-4H2/t6-,7+,8?/m1/s1

InChI key

BMAXQTDMWYDIJX-KVARREAHSA-N

General description

The hydrogenation of 5-norbornene-2-carbonitrile was studied using real-time mass spectrometric analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto A Paz-Schmidt et al.
Analytical chemistry, 81(9), 3665-3668 (2009-04-01)
A new approach for real-time mass spectrometric analysis and identification of intermediates in confined, pressurized vessels is presented. The high-pressure reaction media is depressurized, mixed with a solvent, and fed directly to the atmospheric pressure electrospray ionization source of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service