Skip to Content
Merck
All Photos(1)

Key Documents

14979

Sigma-Aldrich

Boc-β-Homophe-OH

≥98.0% (TLC)

Synonym(s):

(S)-3-(Boc-amino)-4-phenylbutyric acid, Boc-L-β-homophenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
Beilstein:
3060457
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

optical activity

[α]20/D −19.0±2°, c = 1% in methylene chloride

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

101-107 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@H](CC(O)=O)Cc1ccccc1

InChI

1S/C15H21NO4/c1-15(2,3)20-14(19)16-12(10-13(17)18)9-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,16,19)(H,17,18)/t12-/m0/s1

InChI key

ACKWQHCPHJQANL-LBPRGKRZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service