Skip to Content
Merck
All Photos(2)

Key Documents

132276

Sigma-Aldrich

Undecylenic aldehyde

95%

Synonym(s):

10-Undecenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CH(CH2)8CHO
CAS Number:
Molecular Weight:
168.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

95%

form

liquid

refractive index

n20/D 1.4427 (lit.)

bp

98-100 °C/3 mmHg (lit.)

density

0.84 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)CCCCCCCCC=C

InChI

1S/C11H20O/c1-2-3-4-5-6-7-8-9-10-11-12/h2,11H,1,3-10H2

InChI key

OFHHDSQXFXLTKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>140.0 °F - closed cup

Flash Point(C)

> 60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shigeru Kobayashi et al.
Carbohydrate research, 337(11), 1047-1053 (2002-06-01)
The 6-O-mesyl derivative of phenyl 1-thio-beta-D-glucopyranoside was prepared from D-glucose as a synthetic equivalent of a 6-deoxy-hexosyl donor. Racemic methyl 11-hydroxytetradecanoate (methyl convolvulinolate) was synthesized by Grignard reaction of propylmagnesium bromide with 10-undecenal followed by hydroboration. Both intermediates were coupled
Synthesis of new mono and bis amides projected as potential histone deacetylase (HDAC) inhibitors.
Terracciano S, et al.
Tetrahedron, 66(13), 2520-2528 (2010)

Articles

Explore methods for molecular monolayers on silicon surfaces, their properties, and applications in molecular electronics and sensing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service