Skip to Content
Merck
All Photos(1)

Key Documents

132241

Tryptamine hydrochloride

98%

Synonym(s):

2-(3-Indolyl)ethylamine hydrochloride, 3-(2-Aminoethyl)indole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2 · HCl
CAS Number:
Molecular Weight:
196.68
Beilstein:
3568419
EC Number:
MDL number:

Assay

98%

mp

253-255 °C (lit.)

SMILES string

Cl[H].NCCc1c[nH]c2ccccc12

InChI

1S/C10H12N2.ClH/c11-6-5-8-7-12-10-4-2-1-3-9(8)10;/h1-4,7,12H,5-6,11H2;1H

InChI key

KDFBGNBTTMPNIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Tryptamine derivatives as inhibitors against hepatitis B virus
  • Carbamoyl epipodophyllotoxins as potential antitumor agents
  • Anthranilic acid derivatives as CCK receptor antagonists
  • Brassinin derivatives as indoleamine 2,3-dioxygenase inhibitors
  • Antispasmodic agents
  • β-carbolinium cations as new antimalarial agents

Biochem/physiol Actions

Vasoactive; may have a neuromodulator function.

Other Notes

Biogenic amine formed from the decarboxylation of tryptophan by L-aromatic amino acid decarboxylase.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service