Skip to Content
Merck
All Photos(1)

Key Documents

114561

Sigma-Aldrich

Xylenol Blue sodium salt

indicator grade, dye content 90 %

Synonym(s):

p-Xylenolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21NaO5S
CAS Number:
Molecular Weight:
432.46
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
Pricing and availability is not currently available.

grade

indicator grade

composition

dye content, 90%

mp

>300 °C (lit.)

λmax

425 nm in methanol

SMILES string

[Na+].Cc1cc(c(C)cc1O)C(=C2/C=C(C)C(=O)C=C2C)\c3ccccc3S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service