PHL84703
Usaramine N-oxide
phyproof® Reference Substance
Synonym(s):
trans-Retrorsine N-oxide, Mucronatine N-oxide
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
form
solid
manufacturer/tradename
PhytoLab
storage temp.
−20°C
SMILES string
[H][C@]1(C(CO2)=CC3)[N]3(O)CC[C@]1(OC(/C(C[C@@H](C)[C@](O)(CO)C2=O)=C/C)=O)[H]
General description
Check out our entire range of pyrrolizidine alkaloid standards
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service