Skip to Content
Merck
All Photos(1)

Key Documents

G005065

GC Analysis of Ibuprofen Enantiomers (Underivatized) on β-DEX 120

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

column: Supelco β-DEX 120, 30 m x 0.25 mm I.D., 0.25 μm (24304)
gradient: 50 °C (5 min), 5 °C/min to 200 °C (10 min)
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: helium, 60 m/sec., constant flow
injection: 1 μL, 100:1 split
sample: 2 mg/mL in methanol
peak 1: S(+)-ibuprofen
peak 2: R(-)-ibuprofen

liner: 4 mm I.D., FocusLiner

application(s)

clinical
pharmaceutical (small molecule)

Legal Information

DEX is a trademark of Sigma-Aldrich Co. LLC
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.

Analyte

Description

    (S)-(+)-Ibuprofen, solid
    (R)-(−)-Ibuprofen, solid

analytical column

Product No.
Description
Pricing

standard


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service