Skip to Content
Merck
All Photos(11)

Key Documents

ZMS1008

Sigma-Aldrich

Anti-Myc tag Antibody, clone 4A6 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4A6, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

observed mol wt ~55 kDa

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 4A6 is a ZooMAb® mouse recombinant monoclonal antibody that detects c-Myc tagged proteins.

Immunogen

A linear peptide corresponding to amino acids 410-419 from the C-terminal region of human c-Myc.

Application

Quality Control Testing

Evaluated by Western Blotting in Myc-tagged recombinant human p53.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Myc-tagged recombinant human p53.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Myc tag in Myc tag transfected HeLa cells.

Affinity Binding Assay: A representative lot of this antibody bound Myc-tagged p53 protein with a KD of 7.8 x 10-10 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Myc tag, clone 4A6 ZooMAb®, Cat. No. ZMS1008, is a recombinant Mouse monoclonal antibody that detects Myc-tagged proteins and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Western Blotting.

Target description

Identification of the target epitope is important for characterization of a monoclonal antibodies. Epitope tags are short, linear antibody recognition sequences that enable detection of tagged fusion proteins by antibodies. Over the years a wide range of anti-tag antibodies have been developed and each one of these offer unique advantages and disadvantages. Epitope tags can undergo post-translational modifications, such as phosphorylation and glycosylation in cells and cause an increase in the molecular mass of the fused protein that can result in changes in gel mobility. Epitope tags of smaller number of amino acid residues are advantageous in that they exhibit minimal side effects on structure and function of fused target protein. A Myc tag can be used in many different immunoassays and is useful when there is no antibody against the protein of interest. Adding a Myc-tag permits the detection of such proteins with an antibody against the Myc epitope. Clone 4A6 is an antibody that recognizes ten amino acid sequence EQKLISEEDL (Glu-Gln-Lys-Leu-Iso-Ser-Glu-Glu-Asp-Leu) corresponding to the C-terminal region of human c-Myc protein (UniProt: P01106). This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref. Schuchner, S., et al. (2020). Sci. Signal. 13(616); eaax9730; Charest-Morin, X., et al. (2013). Front. Pharmacol. 4: 122).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service