Z722529
X-Pierce™ film
sterile
Synonym(s):
piercable film, piercable plate seal
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Recommended Products
material
vinyl film
sterility
sterile
feature
piercable
manufacturer/tradename
Excel Scientific XP-25
packaging
pkg of one tamper evident zippered bag of 50
technique(s)
tissue processing: suitable
W × L
79.4 mm × 145.5 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Also useful for protecting contents of plate from contamination during multiple manual reagent additions over time. X-Pierce films are 89 μm thick vinyl with a 18 μm thick adhesive layer, designed for temporary protection from contamination and evaporation for samples in 96 well plates. A precut "X" over each well creates four flaps that easily bend downward when pushed by a robotic probe or pipette tip, allowing sample access without coring or adhesive fouling. A thin adhesive layer and special hairline cuts separating the X-Pierce flaps combine to prevent probes from contacting adhesive and becoming occluded. The flaps return to their original position after sampling for continued protection. For long-term sample protection after sampling, a continuous film should be applied as a second layer.
Legal Information
X-Pierce is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service