Z324612
BRAND® SILBERBRAND ETERNA measuring cylinder, low-form
volume 2,000 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
according to DIN ISO 384
feature
subdivision 50 mL
manufacturer/tradename
BRAND
H
340 mm
volume
2,000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Measuring cylinders, low form, short graduation marks. Made from DURAN®, calibrated to contain (TC, In). With spout and hexagonal base. Graduations and inscriptions in ETERNA amber duffusion stain, particularly resistant to aggressive cleaning methods. Tolerances according to DIN EN ISO 4788.
Other Notes
BR42064 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service