Skip to Content
Merck
All Photos(1)

Key Documents

SML2198

Sigma-Aldrich

sAJM589

≥98% (HPLC)

Synonym(s):

5-Hydroxybenzo[a]phenazine, Benzo[a]phenazin-5-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N2O
CAS Number:
Molecular Weight:
246.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

OC1=C2C(C=CC=C2)=C3C(N=C(C=CC=C4)C4=N3)=C1

InChI key

WZBDRAAGHRCRKM-UHFFFAOYSA-N

Biochem/physiol Actions

Myc inhibitor that disrupts the Myc-Max heterodimer
sAJM589 is a Myc inhibitor that disrupts the Myc-Max heterodimer with an IC50 value of 1.8 μM. sAJM589 was shown to suppress cellular proliferation in a variety of MYC-dependent cancer cell line including strong inhibition of Burkitt lymphoma Raji cell line growth. The mechanism of action of sAJM589 is thought to involve increased ubiquitination of Myc by inhibition of Max binding to Myc, leading to proteasome-mediated degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung H Choi et al.
ACS chemical biology, 12(11), 2715-2719 (2017-10-05)
Myc plays important roles in cell cycle progression, cell growth, and stem cell self-renewal. Although dysregulation of Myc expression is a hallmark of human cancers, there is no Myc targeted therapy yet. Here, we report sAJM589, a novel small molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service