Skip to Content
Merck
All Photos(1)

Key Documents

SML0242

Sigma-Aldrich

AZ-MTAB

≥98% (HPLC)

Synonym(s):

3-((1R,3r,5S)-8-((5-Methylthiophen-2-yl)methyl)-8-azabicyclo[3.2.1]octan-3-yloxy)benzamide, 3-[[(3-endo)-8-[(5-methyl-2-thienyl)methyl]-8-azabicyclo[3.2.1]oct-3-yl]oxy]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2S
CAS Number:
Molecular Weight:
356.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL at warmed to 60 °C

storage temp.

2-8°C

SMILES string

Cc1ccc(CN2[C@H]3CC[C@@H]2C[C@@H](C3)Oc4cccc(c4)C(N)=O)s1

InChI

1S/C20H24N2O2S/c1-13-5-8-19(25-13)12-22-15-6-7-16(22)11-18(10-15)24-17-4-2-3-14(9-17)20(21)23/h2-5,8-9,15-16,18H,6-7,10-12H2,1H3,(H2,21,23)/t15-,16+,18+

InChI key

RLHOWHVHHMGIKE-VQFNDLOPSA-N

Biochem/physiol Actions

AZ-MTAB is a potent, short acting κ-opioid receptor (KOR) selective antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to opioid receptors for your research needs.

We offers many products related to opioid receptors for your research needs.

We offers many products related to opioid receptors for your research needs.

We offers many products related to opioid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service